4-Methyl-1,3-dihydro-2H-benzimidazol-2-one 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Methyl-1H-benzo[d]imidazol-2(3H)-one

OR200060-1G OR200060-5G
77406-528EA 95.6 USD
77406-528 77406-530
4-Methyl-1,3-dihydro-2H-benzimidazol-2-one 97%
4-Methyl-1,3-dihydro-2H-benzimidazol-2-one
Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 273…280 °C
Storage Temperature: Ambient
MDL Number: MFCD08274669
CAS Number: 19190-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR