4,5-Dibromo-2-phenylpyridazin-3(2H)-one

Supplier: APOLLO SCIENTIFIC
Warning

OR21545-1G OR21545-250MG
77406-710EA 34.64 USD
77406-710 77406-712
4,5-Dibromo-2-phenylpyridazin-3(2H)-one
4,5-Dibromo-2-phenylpyridazin-3(2H)-one
Formula: C₁₀H₆Br₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00052907
CAS Number: 14305-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR