Methyl-1H-1,2,3-benzotriazole-5-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR26811-25G OR26811-5G
77407-848EA 434.02 USD
77407-848 77407-850
Methyl-1H-1,2,3-benzotriazole-5-carboxylate 98%
Methyl-1H-1,2,3-benzotriazole-5-carboxylate
Formula: C₈H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00174276
CAS Number: 113053-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR