1-(2,4-Dihydroxy-5-propylphenyl)ethanone
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR21981-1G
OR21981-5G
77406-760EA
88.64
USD
77406-760
77406-762
1-(2,4-Dihydroxy-5-propylphenyl)ethanone
1-(2,4-Dihydroxy-5-propylphenyl)ethanone
Formula:
C₁₁H₁₄O₃ Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100627 CAS Number: 63411-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...