5-Isoxazol-5-ylthiophene-2-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR4941-1G OR4941-250MG OR4941-5G
77410-080EA 135.5 USD
77410-080 77410-082 77410-084
5-Isoxazol-5-ylthiophene-2-sulfonyl chloride
5-Isoxazol-5-ylthiophene-2-sulfonyl chloride
Formula: C₇H₄ClNO₃S₂
MW: 249.7 g/mol
Melting Pt: 69…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02187482
CAS Number: 551930-53-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR