Ethyl-2-(bromomethyl)benzoate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethyl 2-(bromomethyl)benzoate

OR6441-1G OR6441-250MG
77410-840EA 98.82 USD
77410-840 77410-842
Ethyl-2-(bromomethyl)benzoate 97%
Ethyl-2-(bromomethyl)benzoate
Formula: C₁₀H₁₁BrO₂
MW: 243.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235262
CAS Number: 7115-91-5
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR