2-Tetrahydro-2H-pyran-4-ylethanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Tetrahydro-2H-pyran-4-yl)ethanol, 4-(2-Hydroxyethyl)tetrahydro-2H-pyran

OR14942-10G OR14942-1G OR14942-5G
77405-198EA 355.56 USD
77405-198 77405-200 77405-202
2-Tetrahydro-2H-pyran-4-ylethanol
2-Tetrahydro-2H-pyran-4-ylethanol
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 119…120 °C/14 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00129068
CAS Number: 4677-18-3
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR