2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR21964-1G OR21964-250MG
77406-752EA 41.78 USD
77406-752 77406-754
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid 95%
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
Formula: C₁₂H₁₁NO₃
MW: 217.23 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00100005
CAS Number: 107367-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR