2,2'-Bipyridine-5-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (2,2-Bipyridine)-5-carboxylic acid

OR18622-100MG OR18622-250MG
77406-368EA 38.72 USD
77406-368 77406-370
2,2'-Bipyridine-5-carboxylic acid 95%
2,2'-Bipyridine-5-carboxylic acid
Formula: C₁₁H₈N₂O₂
MW: 200.2 g/mol
Melting Pt: 196…198 °C
Storage Temperature: Ambient
MDL Number: MFCD08234850
CAS Number: 1970-80-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR