3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR23313-1G OR23313-250MG OR23313-5G
77407-240EA 138.78 USD
77407-240 77407-242 77407-244
3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid 95%
3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid
Formula: C₁₁H₉NO₂S
Melting Pt: 200…201 °C
Storage Temperature: Ambient
MDL Number: MFCD02682065
CAS Number: 28077-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR