3-(4-Methylpiperazin-1-yl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR13862-1G OR13862-25G OR13862-5G
77404-746EA 48.9 USD
77404-746 77404-748 77404-750
3-(4-Methylpiperazin-1-yl)benzoic acid
3-(4-Methylpiperazin-1-yl)benzoic acid
Formula: C₁₂H₁₆N₂O₂
Melting Pt: 187…190 °C
Storage Temperature: Ambient
MDL Number: MFCD06742238
CAS Number: 215309-01-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR