5-Amino-2-nitropyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR18602-250MG OR18602-5G OR18602-1G
77406-354EA 63.16 USD
77406-354 77406-356 77406-352
5-Amino-2-nitropyridine 95%
5-Amino-2-nitropyridine
Formula: C₅H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01646117
CAS Number: 14916-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR