3-(Difluoromethyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC49196-1G PC49196-25G PC49196-5G
77415-888EA 32 USD
77415-888 77415-890 77415-892
3-(Difluoromethyl)benzonitrile
3-(Difluoromethyl)benzonitrile
Formula: C₈H₅F₂N
Storage Temperature: Ambient
MDL Number: MFCD19053256
CAS Number: 55805-13-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR