Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 3-aminothieno[2,3-b]pyridine-2-carboxylate

OR17991-5G OR17991-10G OR17991-1G
77406-174EA 103.92 USD
77406-174 77406-170 77406-172
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Formula: C₉H₈N₂O₂S
MW: 208.24 g/mol
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00174679
CAS Number: 111042-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR