7-Nitro-2,3-dihydro-1-benzofuran-5-sulfonoylchloride

Supplier: APOLLO SCIENTIFIC
Danger

OR103649-1G OR103649-500MG
77402-638EA 176.26 USD
77402-638 77402-640
7-Nitro-2,3-dihydro-1-benzofuran-5-sulfonoylchloride
7-Nitro-2,3-dihydro-1-benzofuran-5-sulfonoylchloride
Formula: C₈H₆ClNO₅S
Melting Pt: 111…113 °C
Storage Temperature: Ambient
MDL Number: MFCD03408011
CAS Number: 874781-18-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR