Tetrafluorosuccinyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC6894-1G PC6894-5G
77417-590EA 156.9 USD
77417-590 77417-592
Tetrafluorosuccinyl chloride 95%
Tetrafluorosuccinyl chloride
Formula: C₄Cl₂F₄O₂
Boiling Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00013653
CAS Number: 356-15-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR