Perfluoro-3,6,9-trioxadecanoic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2972-1G PC2972-25G PC2972-5G
77413-988EA 21.34 USD
77413-988 77413-990 77413-992
Perfluoro-3,6,9-trioxadecanoic acid 98%
Perfluoro-3,6,9-trioxadecanoic acid
Formula: C₇HF₁₃O₅
Boiling Pt: 170…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00156025
CAS Number: 151772-59-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR