5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC0273-1G PC0273-250MG
77412-474EA 227.2 USD
77412-474 77412-476
5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulfonyl chloride 95%
5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulfonyl chloride
Formula: C₉H₆ClF₃N₂O₂S₂
Melting Pt: 65…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00830811
CAS Number: 230295-11-1
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR