3-Fluoro-4-methylphenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC0368-1G PC0368-25G PC0368-5G
77412-524EA 24.6 USD
77412-524 77412-526 77412-528
3-Fluoro-4-methylphenylacetonitrile 98%
3-Fluoro-4-methylphenylacetonitrile
Formula: C₉H₈FN
Storage Temperature: Ambient
MDL Number: MFCD01631535
CAS Number: 261951-73-9
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR