3-Chloro-2-fluoroprop-1-ene 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC3775-1G PC3775-250MG PC3775-5G
77414-878EA 131.42 USD
77414-878 77414-880 77414-882
3-Chloro-2-fluoroprop-1-ene 95%
3-Chloro-2-fluoroprop-1-ene
Formula: C₃H₄ClF
Boiling Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD01696350
CAS Number: 6186-91-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR