2-Oxa-6-azaspiro[3.5]nonane 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR17716-100MG OR17716-250MG
77406-096EA 172.18 USD
77406-096 77406-098
2-Oxa-6-azaspiro[3.5]nonane 95%
2-Oxa-6-azaspiro[3.5]nonane
Formula: C₇H₁₃NO
Boiling Pt: 208 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD12407095
CAS Number: 1046153-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR