(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-2-(hydroxymethyl)-1-methyl-1H-imidazole

OR15479-100MG OR15479-1G OR15479-500MG
77405-414EA 48.9 USD
77405-414 77405-416 77405-418
(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol 97%
(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol
Formula: C₅H₇BrN₂O
MW: 191.03 g/mol
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD09864833
CAS Number: 861362-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR