5-Chloro-6-hydroxynicotinaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR110376-100MG OR110376-1G
77403-290EA 20.28 USD
77403-290 77403-292
5-Chloro-6-hydroxynicotinaldehyde
5-Chloro-6-hydroxynicotinaldehyde
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972384
CAS Number: 627501-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR