Methyl-2-(1,3-benzoxazol-5-yl)acetate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-(benzo[d]oxazol-5-yl)acetate

OR15291-100MG OR15291-500MG
77405-352EA 31.6 USD
77405-352 77405-354
Methyl-2-(1,3-benzoxazol-5-yl)acetate
Methyl-2-(1,3-benzoxazol-5-yl)acetate
Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07021359
CAS Number: 97479-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR