1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Bromoacetyl)benzo[b]thiophene

OR23175-1G OR23175-250MG
77407-160EA 75.4 USD
77407-160 77407-162
1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone 97%
1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone
Formula: C₁₀H₇BrOS
Melting Pt: 138…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00176410
CAS Number: 26167-45-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR