5-(4-Chlorophenyl)-2-thiophenecarbaldehyde

Supplier: APOLLO SCIENTIFIC

OR15397-1G OR15397-5G
77405-390EA 93.74 USD
77405-390 77405-392
5-(4-Chlorophenyl)-2-thiophenecarbaldehyde
5-(4-Chlorophenyl)-2-thiophenecarbaldehyde
Formula: C₁₁H₇ClOS
MW: 222.69 g/mol
Melting Pt: 85…86 °C
Storage Temperature: Ambient
MDL Number: MFCD01860767
CAS Number: 38401-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR