Methyl 6-hydroxy-5-iodonicotinate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 6-hydroxy-5-iodopyridine-3-carboxylate, 2-Hydroxy-3-iodo-5-(methoxycarbonyl)pyridine

OR110402-1G OR110402-250MG OR110402-5G
77403-308EA 29.54 USD
77403-308 77403-310 77403-312
Methyl 6-hydroxy-5-iodonicotinate
Methyl 6-hydroxy-5-iodonicotinate
Formula: C₇H₆INO₃
MW: 279.04 g/mol
Melting Pt: 203…204 °C
Storage Temperature: Ambient
MDL Number: MFCD17168292
CAS Number: 885950-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR