1-(Chloromethyl)-2-(trifluoromethoxy)benzene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Trifluoromethoxy)benzyl chloride

PC7438UE-1G PC7438UE-5G
77417-936EA 28.26 USD
77417-936 77417-938
1-(Chloromethyl)-2-(trifluoromethoxy)benzene 98%
1-(Chloromethyl)-2-(trifluoromethoxy)benzene
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Boiling Pt: 68…70 °C/15 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00221456
CAS Number: 116827-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR