3-(Methylsulfonyl)-1-propanol 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-(Methylsulphonyl)propan-1-ol

OR953133-1G OR953133-5G OR953133-10G
77412-174EA 21.36 USD
77412-174 77412-176 77412-172
3-(Methylsulfonyl)-1-propanol 95%
3-(Methylsulfonyl)-1-propanol
Formula: C₄H₁₀O₃S
MW: 138.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01696727
CAS Number: 2058-49-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR