[2-(4-Aminomethylphenoxy)ethyl]dimethylamine 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-(4-(Aminomethyl)phenoxy)-N,N-dimethylethanamine, 2-[4-(Amino methyl) phenoxy]-n, n-dimethyl ethanamine, Trimethobenzamide Impurity 1

OR345043-1G OR345043-5G
77409-248EA 48.9 USD
77409-248 77409-250
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine 97%
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine
Formula: C₁₁H₁₈N₂O
MW: 194.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075231
CAS Number: 20059-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR