Dimethyl(2-piperazin-1-ylethyl)amine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-[2-(Dimethylamino)ethyl]piperazine

OR6907-1G OR6907-25G OR6907-5G
77410-930EA 69.28 USD
77410-930 77410-932 77410-934
Dimethyl(2-piperazin-1-ylethyl)amine 98%
Dimethyl(2-piperazin-1-ylethyl)amine
Formula: C₈H₁₉N₃
MW: 157.26 g/mol
Boiling Pt: 90 °C/9 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00167969
CAS Number: 3644-18-6
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR