4-Chloro-6-piperidin-1-ylpyrimidin-2-ylamine

Supplier: APOLLO SCIENTIFIC
Warning

OR110706-1G OR110706-250MG OR110706-5G
77403-444EA 220.08 USD
77403-444 77403-446 77403-448
4-Chloro-6-piperidin-1-ylpyrimidin-2-ylamine
4-Chloro-6-piperidin-1-ylpyrimidin-2-ylamine
Formula: C₉H₁₃ClN₄
MW: 212.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664698
CAS Number: 104637-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR