2-(1H-Pyrazol-4-yl)pyrazine

Supplier: APOLLO SCIENTIFIC
Warning

OR1028-1G OR1028-5G
77402-560EA 151.8 USD
77402-560 77402-566
2-(1H-Pyrazol-4-yl)pyrazine
2-(1H-Pyrazol-4-yl)pyrazine
Formula: C₇H₆N₄
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD02579320
CAS Number: 849924-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR