1H,1H-Heptafluorobutyl acrylate 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC4394-1G
PC4394-25G
PC4394-5G
77415-396EA
24.46
USD
77415-396
77415-398
77415-400
1H,1H-Heptafluorobutyl acrylate 97%
1H,1H-Heptafluorobutyl acrylate
Formula:
C₇H₅F₇O₂ MW: 254.11 g/mol Boiling Pt: 120…122 °C/743 mmHg Density: 1.418 g/cm³ (25 °C) Flash Pt: 31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039252 CAS Number: 424-64-6 EINECS: 207-036-8 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...