4-(4,6-Dimethoxypyrimidin-2-yl)aniline 95+%

Supplier: APOLLO SCIENTIFIC
Danger

OR7881-1G OR7881-5G
77411-302EA 198.66 USD
77411-302 77411-304
4-(4,6-Dimethoxypyrimidin-2-yl)aniline 95+%
4-(4,6-Dimethoxypyrimidin-2-yl)aniline
Formula: C₁₂H₁₃N₃O₂
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD01319482
CAS Number: 387350-86-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR