Ethyl-5-nitro-1H-pyrrole-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 5-nitro-1H-pyrrole-2-carboxylate

OR17066-1G OR17066-500MG
77405-946EA 684.64 USD
77405-946 77405-948
Ethyl-5-nitro-1H-pyrrole-2-carboxylate
Ethyl-5-nitro-1H-pyrrole-2-carboxylate
Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Melting Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD09025771
CAS Number: 36131-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR