2-Amino-5-chloro-4-methylthiazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR15552-1G OR15552-5G
77405-442EA 103.92 USD
77405-442 77405-444
2-Amino-5-chloro-4-methylthiazole 95%
2-Amino-5-chloro-4-methylthiazole
Formula: C₄H₅ClN₂S
MW: 148.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699130
CAS Number: 5316-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR