4-(Chloromethyl)-2-isopropylthiazole, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR15567-1G OR15567-250MG OR15567-5G
77405-448EA 82.72 USD
77405-448 77405-450 77405-452
4-(Chloromethyl)-2-isopropylthiazole, Technical Grade
4-(Chloromethyl)-2-isopropylthiazole
Formula: C₇H₁₀ClNS
MW: 175.68 g/mol
Boiling Pt: 54…57 °C/0.07 mmHg
Storage Temperature: Ambient
MDL Number: MFCD06660126
CAS Number: 40516-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR