5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: 5-Amino-1-tert-butyl-1h-pyrazole-4-carbonitrile, 5-Amino-1-(tert-butyl)-1H-pyrazole-4-carbonitrile

OR111100-1G OR111100-5G
77403-646EA 94.76 USD
77403-646 77403-648
5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile 95%
5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile
Formula: C₈H₁₂N₄
MW: 164.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128289
CAS Number: 158001-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR