Methyl amino(3,4-dichlorophenyl)acetate hydrochloride 95%

Supplier: APOLLO SCIENTIFIC

OR12471-1G OR12471-250MG
77404-320EA 428.26 USD
77404-320 77404-322
Methyl amino(3,4-dichlorophenyl)acetate hydrochloride 95%
Methyl amino(3,4-dichlorophenyl)acetate hydrochloride
Formula: C₉H₁₀Cl₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD04115512
CAS Number: 1078611-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR