8-Methyl-4-quinolinol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Hydroxy-8-methylquinoline, 8-Methylquinolin-4-ol

OR12150-1G OR12150-5G
77404-216EA 27.32 USD
77404-216 77404-218
8-Methyl-4-quinolinol
8-Methyl-4-quinolinol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00272344
CAS Number: 23432-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR