6-(Dimethylamino)nicotinonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR12084-1G OR12084-5G
77404-162EA 137.54 USD
77404-162 77404-164
6-(Dimethylamino)nicotinonitrile
6-(Dimethylamino)nicotinonitrile
Formula: C₈H₉N₃
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD08272099
CAS Number: 154924-17-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR