Ethyl-1-(2-formyl-4-nitrophenyl)-4-piperidinecarboxylate

Supplier: APOLLO SCIENTIFIC

OR14119-10G OR14119-1G OR14119-5G
77404-816EA 349.46 USD
77404-816 77404-818 77404-820
Ethyl-1-(2-formyl-4-nitrophenyl)-4-piperidinecarboxylate
Ethyl-1-(2-formyl-4-nitrophenyl)-4-piperidinecarboxylate
Formula: C₁₅H₁₈N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD06770676
CAS Number: 879450-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR