Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 3-bromo-4-hydroxy-5-nitrobenzoate

OR14142-5G OR14142-1G
77404-834EA 131.42 USD
77404-834 77404-832
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 95%
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate
Formula: C₈H₆BrNO₅
MW: 276.05 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD08689690
CAS Number: 40258-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR