2,3-Dihydro-1-benzofuran-5-sulfonamide

Supplier: APOLLO SCIENTIFIC
Warning

OR15079-1G OR15079-250MG OR15079-5G
77405-276EA 124.3 USD
77405-276 77405-278 77405-280
2,3-Dihydro-1-benzofuran-5-sulfonamide
2,3-Dihydro-1-benzofuran-5-sulfonamide
Formula: C₈H₉NO₃S
MW: 199.23 g/mol
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD03069322
CAS Number: 112894-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR