5-(2,5-Dichlorophenyl)-2-furaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR28219-1G OR28219-250MG
77408-042EA 96.78 USD
77408-042 77408-044
5-(2,5-Dichlorophenyl)-2-furaldehyde
5-(2,5-Dichlorophenyl)-2-furaldehyde
Formula: C₁₁H₆Cl₂O₂
MW: 241.08 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00454841
CAS Number: 99113-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR