4-Chloro-6-morpholin-4-ylpyrimidin-2-ylamine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloro-6-morpholinopyrimidin-2-amine

OR111391-1G OR111391-5G
77403-750EA 74.38 USD
77403-750 77403-752
4-Chloro-6-morpholin-4-ylpyrimidin-2-ylamine
4-Chloro-6-morpholin-4-ylpyrimidin-2-ylamine
Formula: C₈H₁₁ClN₄O
MW: 214.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664697
CAS Number: 339016-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR