6-Isothiocyanato-2,3-dihydro-1,4-benzodioxine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,3-Dihydro-1,4-benzodioxin-6-yl isothiocyanate

OR23008-1G OR23008-250MG
77407-010EA 172.18 USD
77407-010 77407-012
6-Isothiocyanato-2,3-dihydro-1,4-benzodioxine 95%
6-Isothiocyanato-2,3-dihydro-1,4-benzodioxine
Formula: C₉H₇NO₂S
MW: 193.23 g/mol
Melting Pt: 64…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00175465
CAS Number: 141492-50-4
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR