3-Acetylamino-4-methoxybenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR322585-1G OR322585-250MG
77409-180EA 143.12 USD
77409-180 77409-182
3-Acetylamino-4-methoxybenzenesulfonyl chloride
3-Acetylamino-4-methoxybenzenesulfonyl chloride
Formula: C₉H₁₀ClNO₄S
Storage Temperature: Ambient
MDL Number: MFCD00129812
CAS Number: 3746-67-6
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR