2-Chloro-N-(3-fluoro-4-methylphenyl)acetamide

Supplier: APOLLO SCIENTIFIC

PC300959-1G PC300959-5G
77414-060EA 30.56 USD
77414-060 77414-062
2-Chloro-N-(3-fluoro-4-methylphenyl)acetamide
2-Chloro-N-(3-fluoro-4-methylphenyl)acetamide
Formula: C₉H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD03152474
CAS Number: 100599-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR